Biotin hydrazide

Lieferant: Apollo Scientific
Achtung

BIB108-100MG
APOSBIB108-100MGEA 21.9 EUR
APOSBIB108-100MG
Biotin hydrazide
Biotin hydrazide

Carbohydrate reactive. Microscopy studies on membranesRef: O'Shannessy,D.J.et al. (1984) Immun.Letters 8,273-277

Formel: C₁₀H₁₈N₄O₂S
Schmelzpunkt: 240…244 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00078532
CAS-Nummer: 66640-86-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR