Biotin-LC-hydrazide

Lieferant: Apollo Scientific
Achtung

Synonyme: N''-Biotinyl-6-aminohexanoylhydrazide, N-(6-Hydrazinyl-6-oxohexyl)biotinamide, N-(6-Hydrazinyl-6-oxohexyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide, Biotinamidocaproyl hydrazine, Biotin LC hydrazide

BIB109-100MG
APOSBIB109-100MGEA 125 EUR
APOSBIB109-100MG
Biotin-LC-hydrazide
Biotin-LC-hydrazide

Carbohydrate reactive -Spacer armMore sensitive than biotin hydrazide.

Formel: C₁₆H₂₉N₅O₃S
Molecular Weight: 371,50 g/mol
Schmelzpunkt: 211 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00077659
CAS-Nummer: 109276-34-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR