Chinidin 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: (+)-Quinidine

BIB1110-5G
APOSBIB1110-5GEA 21.9 EUR
APOSBIB1110-5G
Chinidin 98%
Quinidine
Formel: C20H24N2O2
Molecular Weight: 324,42 g/mol
Schmelzpunkt: 168…172 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00135581
CAS-Nummer: 56-54-2
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR