Big CHAP (N,N-Bis[3-(D-gluconamido)propyl]cholamid)
Lieferant: Apollo Scientific
|
Achtung
|
BIB1157-1G
APOSBIB1157-1GEA
0
EUR
APOSBIB1157-1G
Big CHAP (N,N-Bis[3-(D-gluconamido)propyl]cholamid)
Big CHAP (N,N-Bis[3-(D-gluconamido)propyl]cholamid)
Formel:
C₄₂H₇₅N₃O₁₆ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082541 CAS-Nummer: 86303-22-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...