5-Bromo-4-chloro-3-indolyl acetate

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-4-chloroindoxyl acetate, 5-Bromo-4-chloro-1H-indol-3-yl acetate, BCDA, 5-Bromo-4-chloro-3-indolyl acetate, 3-Acetoxy-4-chloro-5-bromoindole

BIB1170-100MG
APOSBIB1170-100MGEA 26.1 EUR
APOSBIB1170-100MG
5-Bromo-4-chloro-3-indolyl acetate
5-Bromo-4-chloro-3-indolyl acetate
Formel: C₁₀H₇BrClNO₂
Molecular Weight: 288,525 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00037932
CAS-Nummer: 3252-36-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR