5-Bromo-4-chloro-3-indolyl α-D-mannopyranoside

Lieferant: Apollo Scientific
Achtung

Synonyme: X-α-mannoside, (2R,3S,4S,5S,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

BIB1402-250MG
APOSBIB1402-250MGEA 265 EUR
APOSBIB1402-250MG
5-Bromo-4-chloro-3-indolyl α-D-mannopyranoside
5-Bromo-4-chloro-3-indolyl α-D-mannopyranoside
Formel: C₁₄H₁₅BrClNO₆
Molecular Weight: 408,629 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00078951
CAS-Nummer: 125229-64-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR