5-Bromo-3-indolyl nonanoate

Lieferant: Apollo Scientific
Achtung

Synonyme: Blue-nonanoate

BIB1409-250MG
APOSBIB1409-250MGEA 314 EUR
APOSBIB1409-250MG
5-Bromo-3-indolyl nonanoate
5-Bromo-3-indolyl nonanoate
Formel: C₁₇H₂₂BrNO₂
Molecular Weight: 352,266 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD02683905
CAS-Nummer: 133950-70-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR