6-Bromo-2-naphthyl α-D-mannopyranoside

Lieferant: Apollo Scientific
Achtung

BIB1422-100MG
APOSBIB1422-100MGEA 87 EUR
APOSBIB1422-100MG
6-Bromo-2-naphthyl α-D-mannopyranoside
6-Bromo-2-naphthyl α-D-mannopyranoside
Formel: C₁₆H₁₇BrO₆
Molecular Weight: 385,207 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00067621
CAS-Nummer: 28541-84-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR