Phenyl-β-D-thioglucopyranosid

Lieferant: Apollo Scientific
Achtung

Synonyme: (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(phenylthio)tetrahydro-2H-pyran-3,4,5-triol

BIB1429-500MG
APOSBIB1429-500MGEA 102 EUR
APOSBIB1429-500MG
Phenyl-β-D-thioglucopyranosid
Phenyl-β-D-thioglucopyranosid
Formel: C₁₂H₁₆O₅S
Molecular Weight: 272.317 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD11045365
CAS-Nummer: 2936-70-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR