4-Nitrophenyl α-D-xylopyranoside
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2R,3R,4S,5R)-2-(4-Nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol
BIB1435-100MG
BIB1435-250MG
BIB1435-50MG
APOSBIB1435-100MGEA
102
EUR
APOSBIB1435-100MG
APOSBIB1435-250MG
APOSBIB1435-50MG
4-Nitrophenyl α-D-xylopyranoside
4-Nitrophenyl α-D-xylopyranoside
Formel:
C₁₁H₁₃NO₇ Molecular Weight: 271.223 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00067649 CAS-Nummer: 10238-28-5 EINECS: 233-572-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...