A-4501

Lieferant: Apollo Scientific
Achtung

Synonyme: H-β-Ala-AMC·TFA

BIB1437-50MG
APOSBIB1437-50MGEA 52.2 EUR
APOSBIB1437-50MG
A-4501
A-4501
Formel: C₁₅H₁₅F₃N₂O₅
Molecular Weight: 360,289 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00151970
CAS-Nummer: 201847-54-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR