L-Proline 7-amido-4-methylcoumarin hydrobromide salt

Lieferant: Apollo Scientific
Achtung

Synonyme: L-Proline-AMC.HBr

BIB1444-100MG BIB1444-250MG BIB1444-50MG
APOSBIB1444-100MGEA 87 EUR
APOSBIB1444-100MG APOSBIB1444-250MG APOSBIB1444-50MG
L-Proline 7-amido-4-methylcoumarin hydrobromide salt
L-Proline 7-amido-4-methylcoumarin hydrobromide salt
Formel: C₁₅H₁₆N₂O₃·BrH
Molecular Weight: 353.211 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00213795
CAS-Nummer: 115388-93-7

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR