Ac-D-Lys-OH

Lieferant: Apollo Scientific

BIB1449-250MG BIB1449-1G BIB1449-5G
APOSBIB1449-250MGEA 27.7 EUR
APOSBIB1449-250MG APOSBIB1449-1G APOSBIB1449-5G
Ac-D-Lys-OH
Ac-D-Lys-OH
Formel: C₈H₁₆N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00066056
CAS-Nummer: 58840-79-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR