Ac-D-Lys-OH
Lieferant: Apollo Scientific
BIB1449-250MG
BIB1449-1G
BIB1449-5G
APOSBIB1449-250MGEA
27.7
EUR
APOSBIB1449-250MG
APOSBIB1449-1G
APOSBIB1449-5G
Ac-D-Lys-OH
Ac-D-Lys-OH
Formel:
C₈H₁₆N₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066056 CAS-Nummer: 58840-79-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...