6-(Dimethylamino)purin

Lieferant: Apollo Scientific
Achtung

Synonyme: N,N-Dimethyl-7H-purin-6-amine, N6,N6-Dimethyladenine

BIB1476-10G
APOSBIB1476-10GEA 351 EUR
APOSBIB1476-10G
6-(Dimethylamino)purin
6-(Dimethylamino)purine

Inhibitor of protein and cyclin-dependent kinases.

Formel: C₇H₉N₅
Molecular Weight: 163,1799 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00005573
CAS-Nummer: 938-55-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR