β-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate ≥98% (oxidised form)

Lieferant: Apollo Scientific

Synonyme: NADP sodium hydrate, 1-((2R,3R,4S,5R)-5-((((((((2R,3R,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3-hydroxy-4-(phosphonooxy)tetrahydrofuran-2-yl)methoxy)(hydroxy)phosphoryl)oxy)oxidophosphoryl)oxy)methyl)-3,4-dihydroxytetrahydrofuran-2-yl)-3-carbamoylpyridin-1-ium monosodium salt xhydrate

BIB3013-1G
APOSBIB3013-1GEA 43.5 EUR
APOSBIB3013-1G
β-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate ≥98% (oxidised form)
beta-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate
Formel: C₂₁H₂₇N₇NaO₁₇P₃·xH₂O
Schmelzpunkt: 175…178 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00150480
CAS-Nummer: 698999-85-8
EINECS: 214-664-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR