Nicotinamide adenine dinucleotide phosphate (reduced form) tetrasodium salt ≥98%
Lieferant: Apollo Scientific
|
Achtung
|
BIB3014-100MG
BIB3014-500MG
APOSBIB3014-100MGEA
60.9
EUR
APOSBIB3014-100MG
APOSBIB3014-500MG
Nicotinamide adenine dinucleotide phosphate (reduced form) tetrasodium salt ≥98%
Nicotinamide adenine dinucleotide phosphate (reduced form) tetrasodium salt
Formel:
C₂₁H₂₆N₇O₁₇P₃.4Na Molecular Weight: 833.348 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00036263 CAS-Nummer: 2646-71-1 EINECS: 220-163-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...