Brefeldin A

Lieferant: Apollo Scientific
Gefahr

BIB3042-25MG BIB3042-1G BIB3042-50MG
APOSBIB3042-25MGEA 0 EUR
APOSBIB3042-25MG APOSBIB3042-1G APOSBIB3042-50MG
Brefeldin A
Brefeldin A

BFA is a macrolide antiobiotic. Disrupts mechanism of Golgi apparatus.

Formel: C₁₆H₂₄O₄
Schmelzpunkt: 200…205 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00083258
CAS-Nummer: 20350-15-6
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR