Yeast alpha-glucosidase
Lieferant: Apollo Scientific
BIB3066-100KU
APOSBIB3066-100KUEA
654
EUR
APOSBIB3066-100KU
Yeast alpha-glucosidase
Enzyme
From yeast (Saccharomyces cerevisiae). Specific activity: IU/mg protein>50 units. IU per 1mg powder is approximately 70 units.