Aspergillus niger Glucose oxidase
Lieferant: Apollo Scientific
BIB3068-1MU
APOSBIB3068-1MUEA
435
EUR
APOSBIB3068-1MU
Aspergillus niger Glucose oxidase
Enzyme
Lyophilized powder. From Aspergillus niger. Specific activity: IU/mg protein>300 units. IU per 1mg powder is approximately 450 units.