Leuconostoc mesenteroides Sucrose phosphorylase
Lieferant: Apollo Scientific
BIB3100-500U
APOSBIB3100-500UEA 0 EUR
APOSBIB3100-500U
Leuconostoc mesenteroides Sucrose phosphorylase
Enzyme
Lyophilized powder. From Leuconostoc mesenteroides. Specific activity: IU/mg protein>100 units. IU per 1mg powder is approximately 100 units.