5-Bromo-2-methylindole

Lieferant: Apollo Scientific
Achtung

BIB6015-1G
APOSBIB6015-1GEA 40.6 EUR
APOSBIB6015-1G
5-Bromo-2-methylindole
5-Bromo-2-methylindole
Formel: C₉H₈BrN
Molecular Weight: 210,074 g/mol
Schmelzpunkt: 104…107 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD01863677
CAS-Nummer: 1075-34-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR