5-Brom-DL-tryptophan
Lieferant: Apollo Scientific
Synonyme:
2-Amino-3-(5-bromo-1H-indol-3-yl)propanoic acid
BIB6018-1G
APOSBIB6018-1GEA
162
EUR
APOSBIB6018-1G
5-Brom-DL-tryptophan
5-Brom-DL-tryptophan
Formel:
C₁₁H₁₁BrN₂O₂ Molecular Weight: 283.12 g/mol Schmelzpunkt: 264 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005648 CAS-Nummer: 6548-09-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...