Chlorogenic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: (1S,3R,4R,5R)-3-(((3-(3,4-dihydroxyphenyl)acryloyl)oxy)-1,4,5-trihydroxycyclohexanecarboxylic acid, 3-O-Caffeoylquinic acid, 3-(3,4-Dihydroxycinnamoyl)quinic acid, (1S,3R,4R,5R)-3-[[3-(3,4-Dihydroxyphenyl)-1-oxo-2-propenyl]oxy]-1,4,5-trihydroxycyclohexanecarboxylic acid

BIB6027-25G BIB6027-1G BIB6027-5G
APOSBIB6027-25GEA 155 EUR
APOSBIB6027-25G APOSBIB6027-1G APOSBIB6027-5G
Chlorogenic acid
Chlorogenic acid
Formel: C₁₆H₁₈O₉
Molecular Weight: 354.31 g/mol
Schmelzpunkt: 210 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003862
CAS-Nummer: 327-97-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR