5-Chlor-3-indolylessigsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Chloroindole-3-acetic acid, 2-(5-Chloro-1H-indol-3-yl)acetic acid

BIB6029-500MG
APOSBIB6029-500MGEA 39.3 EUR
APOSBIB6029-500MG
5-Chlor-3-indolylessigsäure
5-Chlor-3-indolylessigsäure
Formel: C₁₀H₈ClNO₂
Molecular Weight: 209,63 g/mol
Schmelzpunkt: 156…160 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00130159
CAS-Nummer: 1912-45-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR