Cholesteryl phosphorylethanolamine

Lieferant: Apollo Scientific

BIB6033-100MG
APOSBIB6033-100MGEA 618 EUR
APOSBIB6033-100MG
Cholesteryl phosphorylethanolamine
Cholesteryl phosphorylethanolamine
Formel: C₂₉H₅₂NO₄P
Molecular Weight: 509,701 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD09841711
CAS-Nummer: 120090-10-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR