α-Cyclodextrin

Lieferant: Apollo Scientific
Achtung

BIB6038-25G
APOSBIB6038-25GEA 21.9 EUR
APOSBIB6038-25G
α-Cyclodextrin
α-Cyclodextrin
Formel: C₃₆H₆₀O₃₀
Molecular Weight: 972,84 g/mol
Schmelzpunkt: >278 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00078207
CAS-Nummer: 10016-20-3
EINECS: 233-007-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR