D(+)-Fucose

Lieferant: Apollo Scientific

BIB6052-1G
APOSBIB6052-1GEA 108 EUR
APOSBIB6052-1G
D(+)-Fucose
D(+)-Fucose
Formel: C₆H₁₂O₅
Schmelzpunkt: 144…145 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00135603
CAS-Nummer: 3615-37-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR