L-Homocysteine

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-2-Amino-4-mercaptobutanoic acid, H-HoCys-OH

BIB6065-100MG BIB6065-1G BIB6065-500MG
APOSBIB6065-100MGEA 109 EUR
APOSBIB6065-100MG APOSBIB6065-1G APOSBIB6065-500MG
L-Homocysteine
L-Homocysteine
Formel: C₄H₉NO₂S
Molecular Weight: 135.19 g/mol
Schmelzpunkt: 263…265 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00151320
CAS-Nummer: 6027-13-0
EINECS: 227-891-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR