Indole-3-pyruvic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: alpha-Oxo-1H-indole-3-propanoic acid, 1H-Indole-3-propanoic acid, .alpha.-oxo-, 3-(1H-Indol-3-yl)-2-oxopropanoic acid, 3-(3-Indolyl)-2-oxopropanoic acid, 3-Indolylpyruvic acid

BIB6079-1G
APOSBIB6079-1GEA 277 EUR
APOSBIB6079-1G
Indole-3-pyruvic acid
Indole-3-pyruvic acid
Formel: C₁₁H₉NO₃
Molecular Weight: 203.197 g/mol
Schmelzpunkt: 215 °C (Zers.)
Lagertemperatur: Gefrierschrank
MDL: MFCD00005640
CAS-Nummer: 392-12-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR