Isopropyl-β-D-thioglucuronic acid sodium salt
Lieferant: Apollo Scientific
|
Achtung
|
BIB6085-250MG
APOSBIB6085-250MGEA
212
EUR
APOSBIB6085-250MG
Isopropyl-β-D-thioglucuronic acid sodium salt
Isopropyl-β-D-thioglucuronic acid sodium salt
Formel:
C₉H₁₅O₆S·Na Lagertemperatur: Gefrierschrank |
MDL:
MFCD00798401 CAS-Nummer: 208589-93-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...