D-Kynurenine free base

Lieferant: Apollo Scientific
Achtung

Synonyme: β-Anthraniloyl-D-alanine, (R)-2-Amino-4-(2-aminophenyl)-4-oxobutanoic acid, D-Kynurenine

BIB6087-1G
APOSBIB6087-1GEA 837 EUR
APOSBIB6087-1G
D-Kynurenine free base
D-Kynurenine free base
Formel: C₁₀H₁₂N₂O₃
Molecular Weight: 208.214 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00133224
CAS-Nummer: 13441-51-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR