DL-Lauroylcarnitine chloride

Lieferant: Apollo Scientific
Achtung

Synonyme: Lauroyl-DL-carnitine chloride

BIB6090-1G
APOSBIB6090-1GEA 306 EUR
APOSBIB6090-1G
DL-Lauroylcarnitine chloride
DL-Lauroylcarnitine chloride
Lagertemperatur: Gefrierschrank
MDL: MFCD00063478
CAS-Nummer: 14919-37-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR