L-Lysine 4-nitroanilide dihydrobromide

Lieferant: Apollo Scientific

BIB6094-5G
APOSBIB6094-5GEA 841 EUR
APOSBIB6094-5G
L-Lysine 4-nitroanilide dihydrobromide
L-Lysine 4-nitroanilide dihydrobromide
Formel: C₁₂H₂₀Br₂N₄O₃
Lagertemperatur: Kühlschrank
MDL: MFCD00050686
CAS-Nummer: 40492-96-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR