Methyl-α-D-galactopyranoside
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol, Methyl α-D-galactoside
BIB6104-25G
APOSBIB6104-25GEA
50.9
EUR
APOSBIB6104-25G
Methyl-α-D-galactopyranoside
Methyl-α-D-galactopyranoside
Formel:
C₇H₁₄O₆ Molecular Weight: 194,18 g/mol Schmelzpunkt: 116…117 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00064085 CAS-Nummer: 3396-99-4 EINECS: 222-251-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...