Methyl-α-D-glucopyranosid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
α-D-Methylglucoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol
BIB6105-500G
APOSBIB6105-500GEA
37.7
EUR
APOSBIB6105-500G
Methyl-α-D-glucopyranosid
Methyl-α-D-glucopyranosid
Formel:
C₇H₁₄O₆ Molecular Weight: 194,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064086 CAS-Nummer: 97-30-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...