5-Methyl-DL-tryptophan

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-3-(5-methyl-1H-indol-3-yl)propanoic acid, H-DL-Trp(5-Me)-OH

BIB6112-1G BIB6112-500MG BIB6112-5G
APOSBIB6112-1GEA 261 EUR
APOSBIB6112-1G APOSBIB6112-500MG APOSBIB6112-5G
5-Methyl-DL-tryptophan
5-Methyl-DL-tryptophan
Formel: C₁₂H₁₄N₂O₂
Molecular Weight: 218.26 g/mol
Schmelzpunkt: 280…282 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00005652
CAS-Nummer: 951-55-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR