6-Methyl-DL-tryptophan

Lieferant: Apollo Scientific
Achtung

BIB6113-1G
APOSBIB6113-1GEA 644 EUR
APOSBIB6113-1G
6-Methyl-DL-tryptophan
6-Methyl-DL-tryptophan
Schmelzpunkt: 290 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00022772
CAS-Nummer: 2280-85-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR