4-Nitrophenyl-β-D-fucopyranoside

Lieferant: Apollo Scientific
Achtung

BIB6126-250MG
APOSBIB6126-250MGEA 134 EUR
APOSBIB6126-250MG
4-Nitrophenyl-β-D-fucopyranoside
4-Nitrophenyl-beta-D-fucopyranoside
Formel: C₁₂H₁₅NO₇
Molecular Weight: 285,25 g/mol
Schmelzpunkt: 184 °C
Lagertemperatur: Gefrierschrank
CAS-Nummer: 1226-39-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR