Phenylethyl-β-D-thiogalactopyranoside

Lieferant: Apollo Scientific
Achtung

Synonyme: (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(phenethylthio)tetrahydro-2H-pyran-3,4,5-triol

BIB6257-1G
APOSBIB6257-1GEA 393 EUR
APOSBIB6257-1G
Phenylethyl-β-D-thiogalactopyranoside
Phenylethyl-β-D-thiogalactopyranoside
Formel: C₁₄H₂₀O₅S
Molecular Weight: 300,3706 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00467904
CAS-Nummer: 63407-54-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR