6-TAMRA

Lieferant: Apollo Scientific
Achtung

BIB6288-250MG BIB6288-50MG BIB6288-100MG
APOSBIB6288-250MGEA 0 EUR
APOSBIB6288-250MG APOSBIB6288-50MG APOSBIB6288-100MG
6-TAMRA
6-TAMRA
Formel: C₂₅H₂₂N₂O₅
Lagertemperatur: Raumtemperatur
MDL: MFCD01073638
CAS-Nummer: 91809-67-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR