Atractyloside potassium salt

Lieferant: Apollo Scientific
Gefahr

BIB6292-100MG BIB6292-25MG BIB6292-50MG
APOSBIB6292-100MGEA 641 EUR
APOSBIB6292-100MG APOSBIB6292-25MG APOSBIB6292-50MG
Atractyloside potassium salt
Atractyloside potassium salt
Formel: C₃₀H₄₄K₂O₁₆S₂
Schmelzpunkt: 174 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00078810
CAS-Nummer: 102130-43-8
UN: 2811
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR