Bisacrylamid ≥98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N,N'-Methylendiacrylamid, BIS
BIB6418-100G
BIB6418-1KG
BIB6418-250G
APOSBIB6418-100GEA
71.2
EUR
APOSBIB6418-100G
APOSBIB6418-1KG
APOSBIB6418-250G
Bisacrylamid ≥98%
Bisacrylamid
Formel:
(H₂C=CHCONH)₂CH₂ Schmelzpunkt: >300 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00008625 CAS-Nummer: 110-26-9 EINECS: 203-750-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...