AC1NUZLU

Lieferant: Apollo Scientific
Achtung

Synonyme: Z-Val-Ala-DL-Asp-fluoromethylketone

BIBA1001-5MG
APOSBIBA1001-5MGEA 522 EUR
APOSBIBA1001-5MG
AC1NUZLU
AC1NUZLU
Formel: C₂₁H₂₈FN₃O₇
Molecular Weight: 453,467 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD06795862
CAS-Nummer: 220644-02-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR