Poly(L-Lactide-co-glycolide)
Lieferant: Apollo Scientific
BIBE1001-5G
APOSBIBE1001-5GEA
190
EUR
APOSBIBE1001-5G
Poly(L-Lactide-co-glycolide)
Poly(L-Lactide-co-glycolide)
Polymer. Lactide:glycolide 65:35, viscosity 0.6 dL/g