Poly(caprolactone-co-glycolide) 95:5
Lieferant: Apollo Scientific
BIBE1002-5G
APOSBIBE1002-5GEA 63.8 EUR
APOSBIBE1002-5G
Poly(caprolactone-co-glycolide) 95:5
Poly(caprolactone-co-glycolide)
Polymer. Caprolactone:glycolide 95:5. Viscosity 1.0 dL/g