Poly(caprolactone-co-glycolide) 90:10
Lieferant: Apollo Scientific
BIBE1003-5G
APOSBIBE1003-5GEA
63.8
EUR
APOSBIBE1003-5G
Poly(caprolactone-co-glycolide) 90:10
Poly(caprolactone-co-glycolide)
Polymer. Caprolactone:glycolide 90:10. Viscosity 0.75 dL/g