Benzyl N-(2-aminoethyl)carbamate hydrochloride ≥99%
Lieferant: Apollo Scientific
BIBP1042-5G
APOSBIBP1042-5GEA
171
EUR
APOSBIBP1042-5G
Benzyl N-(2-aminoethyl)carbamate hydrochloride ≥99%
Benzyl N-(2-aminoethyl)carbamate hydrochloride
Formel:
C₁₀H₁₄N₂O₂ Schmelzpunkt: 169…172 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00270150 CAS-Nummer: 18807-71-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...