Caerulomycin A

Lieferant: Apollo Scientific
Gefahr

Synonyme: (E)-4-Methoxy-[2,2'-bipyridine]-6-carbaldehyde oxime

BIBR1019-1MG
APOSBIBR1019-1MGEA 783 EUR
APOSBIBR1019-1MG
Caerulomycin A
Caerulomycin A
Formel: C₁₂H₁₁N₃O₂
Molecular Weight: 229,235 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD03093715
CAS-Nummer: 21802-37-9
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR