Roquefortine C

Lieferant: Apollo Scientific
Gefahr

BIBR1088-1MG
APOSBIBR1088-1MGEA 296 EUR
APOSBIBR1088-1MG
Roquefortine C
Roquefortine C
Formel: C₂₂H₂₃N₅O₂
Lagertemperatur: Gefrierschrank
CAS-Nummer: 58735-64-1
UN: 2811
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR